Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RCC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544747
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544747 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544747 Supplier Invitrogen™ Supplier No. PA544747
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: RIRSLACGKS SIIVAADEST ISWGPSPTFG ELGYGDHKPK SSTAAQEVKT Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.

Required for completion of mitosis and cytokinesis. May function as a guanine nucleotide exchange factor for the small GTPase RAC1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RCC2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RCC2
Gene Accession No. Q8BK67, Q9P258
Gene Alias 2610510H01Rik; 2610529N02Rik; AA536646; AA675016; KIAA1470; mKIAA1470; protein RCC2; Protein RCC2 homolog; RCC1-like; RCC1-like protein TD-60; RCC2; regulator of chromosome condensation 2; RGD1309986; Td60; td-60; Telophase disk protein of 60 kDa; wu:fb36a03; zgc:77115
Gene Symbols RCC2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human RCC2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108911, 55920
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.