Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RCCD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110283
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110283 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110283 Supplier Invitrogen™ Supplier No. PA5110283
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DIYIWGWNES GQLALPTRNL AEDGETVARE ATELNEDGSQ VKRTGGAEDG APAPFIAVQP FPALLDLPMG SDAVKASCGS RHTAVVTRTG ELYTWGWGKY GQLGHEDTTS LDRPRRVEYF VDKQLQVKAV T.

Plays a role in transcriptional repression of satellite repeats, possibly by regulating H3K36 methylation levels in centromeric regions together with KDM8 (PubMed:24981860). Possibly together with KDM8, is involved in proper mitotic spindle organization and chromosome segregation (PubMed:24981860). Plays a role in regulating alpha-tubulin deacetylation and cytoskeletal microtubule stability, thereby promoting cell migration and TGF-beta-induced epithelial to mesenchymal transition (EMT), potentially through the inhibition of KDM8 (PubMed:28455245).
TRUSTED_SUSTAINABILITY

Specifications

Antigen RCCD1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.14 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RCCD1
Gene Accession No. A6NED2
Gene Alias RCC1 domain containing 1; RCC1 domain-containing protein 1; RCCD1
Gene Symbols RCCD1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 230-360 of human RCCD1 (NP_001017919.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 308760, 91433
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.