Learn More
RCD8 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300746A
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse and Rat
In the process of mRNA degradation, seems to play a role in mRNA decapping. Component of a complex containing DCP2 and DCP1A which functions in decapping of ARE-containing mRNAs. Promotes complex formation between DCP1A and DCP2. Enhances the catalytic activity of DCP2 (in vitro).Specifications
| RCD8 | |
| Polyclonal | |
| Unconjugated | |
| EDC4 | |
| Ge-1, HEDLS, RCD-8, autoantigen Ge-1, autoantigen RCD-8, enhancer of mRNA-decapping protein 4, human enhancer of decapping large subunit | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23644 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q6P2E9 | |
| EDC4 | |
| Between 1351 and 1401 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.