Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RCE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599076
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599076 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599076 Supplier Invitrogen™ Supplier No. PA599076
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RCE1 Polyclonal Antibody for Western Blot, ELISA

This gene encodes an integral membrane protein which is classified as a member of the metalloproteinase family. This enzyme is thought to function in the maintenance and processing of CAAX-type prenylated proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RCE1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene RCE1
Gene Accession No. Q9Y256
Gene Alias CAAX prenyl protease 2; CAAX prenyl protein protease RCE1; D19Ertd283e; D19Ertd98e; FACE2; FACE-2; farnesylated protein-converting enzyme 2; Farnesylated proteins-converting enzyme 2; hRCE1; prenyl protein peptidase RCE1; prenyl protein-specific endoprotease 2; Ras and a-factor-converting enzyme 1 homolog; Ras converting CAAX endopeptidase 1; Rce1; RCE1 homolog; RCE1 homolog, prenyl protein peptidase; RCE1 homolog, prenyl protein peptidase (S. cerevisiae); RCE1 homolog, prenyl protein protease; RCE1A; RCE1B
Gene Symbols RCE1
Host Species Rabbit
Immunogen Recombinant Human CAAX prenyl protease 2 protein (133-185AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9986
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.