Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RCE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140156
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140156 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140156 Supplier Invitrogen™ Supplier No. PA5140156
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: LTDPKLYGSL PLCVLLERAG DSEAPLCS Highest antigen sequence indentity to the following orthologs: Mouse - 82%, Rat - 82%.

This gene encodes an integral membrane protein which is classified as a member of the metalloproteinase family. This enzyme is thought to function in the maintenance and processing of CAAX-type prenylated proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RCE1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RCE1
Gene Accession No. Q9Y256
Gene Alias CAAX prenyl protease 2; CAAX prenyl protein protease RCE1; D19Ertd283e; D19Ertd98e; FACE2; FACE-2; farnesylated protein-converting enzyme 2; Farnesylated proteins-converting enzyme 2; hRCE1; prenyl protein peptidase RCE1; prenyl protein-specific endoprotease 2; Ras and a-factor-converting enzyme 1 homolog; Ras converting CAAX endopeptidase 1; Rce1; RCE1 homolog; RCE1 homolog, prenyl protein peptidase; RCE1 homolog, prenyl protein peptidase (S. cerevisiae); RCE1 homolog, prenyl protein protease; RCE1A; RCE1B
Gene Symbols RCE1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RCE1. Recombinant protein control fragment (Product #RP-109390).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9986
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.