Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RCE1 Rabbit anti-Human, Polyclonal, FabGennix

Catalog No. 502208831
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-8831 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-8831 Supplier Fabgennix Inc Supplier No. RCE1101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RCE1 Polyclonal antibody specifically detects RCE1 in Human samples. It is validated for ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen RCE1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene RCE1
Gene Accession No. Q9Y256
Gene Alias CAAX prenyl protease 2; CAAX prenyl protein protease RCE1; D19Ertd283e; D19Ertd98e; FACE2; FACE-2; farnesylated protein-converting enzyme 2; Farnesylated proteins-converting enzyme 2; hRCE1; prenyl protein peptidase RCE1; prenyl protein-specific endoprotease 2; Ras and a-factor-converting enzyme 1 homolog; Ras converting CAAX endopeptidase 1; Rce1; RCE1 homolog; RCE1 homolog, prenyl protein peptidase; RCE1 homolog, prenyl protein peptidase (S. cerevisiae); RCE1 homolog, prenyl protein protease; RCE1A; RCE1B
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid sequence 300-350 on human RCE1 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9986
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.