Learn More
RCL Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304681A

Description
The recommended shelf life for this product is 1 year from date of receipt.
RCL was identified on the basis of its stimulation by c-Myc protein. The latter is a transcription factor that participates in the regulation of cell proliferation, differentiation, and apoptosis. The exact function of this gene is not known but studies in rat suggest a role in cellular proliferation and c-Myc-mediated transformation.Specifications
RCL | |
Polyclonal | |
Unconjugated | |
DNPH1 | |
RCL, RP3-330M21.3, dJ330M21.3, c-Myc-responsive protein Rcl, deoxyribonucleoside 5′-monophosphate N-glycosidase, putative c-Myc-responsive | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10591 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
O43598 | |
DNPH1 | |
Between 124 and 174 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.