Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RCL Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988046
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8046 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8046 Supplier Bioss Supplier No. BS7827R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RCL was identified on the basis of its stimulation by c-Myc protein. The latter is a transcription factor that participates in the regulation of cell proliferation, differentiation, and apoptosis. The exact function of this gene is not known but studies in rat suggest a role in cellular proliferation and c-Myc-mediated transformation.

Specifications

Antigen RCL
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DNPH1
Gene Accession No. O43598
Gene Alias 2′-deoxynucleoside 5′-phosphate N-hydrolase 1; C6orf108; c-Myc-responsive protein RCL; deoxyribonucleoside 5′-monophosphate N-glycosidase; dJ330M21.3; DNPH1; putative c-Myc-responsive; RCL; RP3-330M21.3
Gene Symbols DNPH1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human RCL/c Myc responsive.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10591
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.