Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RCL Recombinant Rabbit Monoclonal Antibody (7C1Z8)
GREENER_CHOICE

Catalog No. PIMA542778
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542778 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542778 Supplier Invitrogen™ Supplier No. MA542778
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: 293T, BT-474. Immunogen sequence: LIHEQDLEWL QQADVVVAEV TQPSLGVGYE LGRAVAFNKR ILCLFRPQSG RVLSAMIRGA ADGSRFQVWD YEEGEVEALL DRYFEADPPG QVAASPDPTT.

RCL was identified on the basis of its stimulation by c-Myc protein. The latter is a transcription factor that participates in the regulation of cell proliferation, differentiation, and apoptosis. The exact function of this gene is not known but studies in rat suggest a role in cellular proliferation and c-Myc-mediated transformation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RCL
Applications ELISA, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 7C1Z8
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DNPH1
Gene Accession No. O43598, Q80VJ3
Gene Alias 2'-deoxynucleoside 5'-phosphate N-hydrolase 1; 2'-deoxynucleoside 5'-phosphate N-hydrolase 1 {ECO:0000255; C6orf108; C76683; c-Myc-responsive protein RCL; c-Myc-responsive protein Rcl {ECO:0000255; Deoxyribonucleoside 5'-monophosphate N-glycosidase; dJ330M21.3; DNPH1; HAMAP-Rule:MF_03036}; Nucleoside 2-deoxyribosyltransferase domain containing protein RGD620382; putative c-Myc-responsive; Rcl; RGD620382; RP3-330M21.3
Gene Symbols DNPH1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 75-174 of human DNPH1 (O43598).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10591, 381101
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.