Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RCL Recombinant Rabbit Monoclonal Antibody (JE59-29)
GREENER_CHOICE

Catalog No. PIMA544893
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544893 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544893 Supplier Invitrogen™ Supplier No. MA544893
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

RCL Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

RCL was identified on the basis of its stimulation by c-Myc protein. The latter is a transcription factor that participates in the regulation of cell proliferation, differentiation, and apoptosis. The exact function of this gene is not known but studies in rat suggest a role in cellular proliferation and c-Myc-mediated transformation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RCL
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JE59-29
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene DNPH1
Gene Accession No. O43598
Gene Alias 2'-deoxynucleoside 5'-phosphate N-hydrolase 1; 2'-deoxynucleoside 5'-phosphate N-hydrolase 1 {ECO:0000255; C6orf108; C76683; c-Myc-responsive protein RCL; c-Myc-responsive protein Rcl {ECO:0000255; Deoxyribonucleoside 5'-monophosphate N-glycosidase; dJ330M21.3; DNPH1; HAMAP-Rule:MF_03036}; Nucleoside 2-deoxyribosyltransferase domain containing protein RGD620382; putative c-Myc-responsive; Rcl; RGD620382; RP3-330M21.3
Gene Symbols DNPH1
Host Species Rabbit
Immunogen Recombinant protein within human RCL aa 75-174/174.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10591
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.