Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RCL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567163
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567163 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567163 Supplier Invitrogen™ Supplier No. PA567163
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RGIGYYLESLL CLAPFMKHPL KIVLRGVTND QVDPSVDVLK ATALPLLKQF GIDGESFELK IVRRGMPPGG GGEVVFSCPV RKVLKPIQLT DPGK Highest antigen sequence identity to the following orthologs - mouse 96%, rat 96%.

RCL1, which does not have cyclase activity, plays a role in 40S- ribosomal-subunit biogenesis in the early pre-rRNA processing steps at sites A0, A1 and A2 that are required for proper maturation of the 18S RNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RCL1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RCL1
Gene Accession No. Q9Y2P8
Gene Alias 2310040A02Rik; Ab1-353; Ac1292; AI789745; C76567; HSPC338; Rcl1; RNA 3'-terminal phosphate cyclase-like protein; RNA cyclase homolog; RNA terminal phosphate cyclase like 1; RNA terminal phosphate cyclase-like 1; Rnac; RPC2; RPCL1; RTC2
Gene Symbols RCL1
Host Species Rabbit
Immunogen Recombinant Human RCL1. Recombinant protein control fragment (Product #RP-105528).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10171
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.