Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RCN1 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB500256JF646
Description
RCN1 Polyclonal antibody specifically detects RCN1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
RCN1 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
FLJ37041, FLJ55835, PIG20, proliferation-inducing gene 20, RCAL, RCN, reticulocalbin 1, EF-hand calcium binding domain, reticulocalbin-1 | |
A synthetic peptide that maps to a region between residues 1 and 75 of human Reticulocalbin-1, EF-hand calcium binding domain using the numbering given in Swiss-Prot entry Q15293 (GeneID 5954). | |
0.1 mL | |
Apoptosis, Signal Transduction, Stem Cell Markers | |
5954 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction