Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RCN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587992
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587992 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587992 Supplier Invitrogen™ Supplier No. PA587992
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KPTVRKERVV RPDSELGERP PEDNQSFQYD HEAFLGKEDS KTFDQLTPDE SKERLGKIVD RIDNDGDGFV TTEELKTWIK RVQKRYIFDN VAKVWKDYDR DKDDKISWEE YKQATYGYYL GNPAEFHDSS DHHTFKKMLP RDERRFKAAD LNGDLTATRE EFTAFLHPEE FEHMKEIVVL ETLEDIDKNG DGFVDQDEYI ADMFSHEENG PEPDWVLSER EQFNEFRDLN KDGKLDKDEI RHWILPQDYD HAQAEARHLV YESDKNKDEK LTKEEILENW NMFVGSQATN YGEDLTKNHD EL; Positive Samples: A-549, SKOV3, LO2, HeLa; Cellular Location: Endoplasmic reticulum lumen.

Reticulocalbin 1 is a calcium-binding protein located in the lumen of the ER. The protein contains six conserved regions with similarity to a high affinity Ca(+2)-binding motif, the EF-hand. High conservation of amino acid residues outside of these motifs, in comparison to mouse reticulocalbin, is consistent with a possible biochemical function besides that of calcium binding. In human endothelial and prostate cancer cell lines this protein localizes to the plasma membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RCN1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RCN1
Gene Accession No. Q05186, Q15293
Gene Alias epididymis secretory protein Li 84; FLJ37041; HEL-S-84; PIG20; proliferation-inducing gene 20; Rca1; RCAL; Rcn; RCN1; reticulocalbin 1; reticulocalbin 1, EF-hand calcium binding domain; reticulocalbin-1
Gene Symbols RCN1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 30-331 of human RCN1 (NP_0028921).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19672, 5954
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.