Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RCN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558409
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558409 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558409 Supplier Invitrogen™ Supplier No. PA558409
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LGKIVDRIDN DGDGFVTTEE LKTWIKRVQK RYIFDNVAKV WKDYDRDKDD KISWEEYKQA Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.

Reticulocalbin 1 is a calcium-binding protein located in the lumen of the ER. The protein contains six conserved regions with similarity to a high affinity Ca(+2)-binding motif, the EF-hand. High conservation of amino acid residues outside of these motifs, in comparison to mouse reticulocalbin, is consistent with a possible biochemical function besides that of calcium binding. In human endothelial and prostate cancer cell lines this protein localizes to the plasma membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RCN1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RCN1
Gene Accession No. Q05186, Q15293
Gene Alias epididymis secretory protein Li 84; FLJ37041; HEL-S-84; PIG20; proliferation-inducing gene 20; Rca1; RCAL; Rcn; RCN1; reticulocalbin 1; reticulocalbin 1, EF-hand calcium binding domain; reticulocalbin-1
Gene Symbols RCN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RCN1. Recombinant protein control fragment (Product #RP-97267).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19672, 5954
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.