Learn More
Invitrogen™ RCN2 Polyclonal Antibody

Description
Immunogen sequence: AEELHYPLGE RRSDYDREAL LGVQEDVDEY VKLGHEEQQK RLQAIIKKID LDSDGFLTES ELSSWIQMSF KHYAMQEAKQ QFVEYDK Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 90%.
Specifications
Specifications
| Antigen | RCN2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | RCN2 |
| Gene Accession No. | Q14257 |
| Gene Alias | AA408742; Calcium-binding protein ERC-55; E6-binding protein; E6BP; ERC55; ERC-55; RCN2; reticulocalbin 2; reticulocalbin 2, EF-hand calcium binding domain; reticulocalbin 2, EF-hand calcium binding domain (endoplasmic reticulum calcium-binding protein, 55kD); reticulocalbin-2; taipoxin-associated calcium binding protein 49; taipoxin-associated calcium-binding protein 49; TCBP49; TCBP-49 |
| Gene Symbols | RCN2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.