Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RCN3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560252
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560252 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560252 Supplier Invitrogen™ Supplier No. PA560252
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DGWVSLAELR AWIAHTQQRH IRDSVSAAWD TYDTDRDGRV GWEELRNATY GHYAPGEEFH DVEDAETYK Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

Probable molecular chaperone assisting protein biosynthesis and transport in the endoplasmic reticulum (PubMed:16433634, PubMed:28939891). Required for the proper biosynthesis and transport of pulmonary surfactant-associated protein A/SP-A, pulmonary surfactant-associated protein D/SP-D and the lipid transporter ABCA3. By regulating both the proper expression and the degradation through the endoplasmic reticulum-associated protein degradation pathway of these proteins plays a crucial role in pulmonary surfactant homeostasis. Has an anti-fibrotic activity by negatively regulating the secretion of type I and type III collagens (PubMed:28939891). This calcium-binding protein also transiently associates with immature PCSK6 and regulates its secretion (PubMed:16433634). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RCN3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RCN3
Gene Accession No. Q96D15
Gene Alias 6030455P07Rik; D530026G20Rik; D7Ertd671e; EF-hand calcium-binding protein RLP49; Rcn3; reticulocabin; reticulocalbin 3; reticulocalbin 3, EF-hand calcium binding domain; reticulocalbin-3; RLP49; UNQ239/PRO272
Gene Symbols RCN3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RCN3. Recombinant protein control fragment (Product #RP-99107).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57333
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.