Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RCOR1/CoREST Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$382.00 - $610.00

Specifications

Antigen RCOR1/CoREST
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP238720
SDP
View Documents
Novus Biologicals
NBP238720
0.1 mL
Each for $610.00
Only null left
Add to Cart
 
NB396699
SDP
View Documents
Novus Biologicals
NBP23872025UL
25 μL
Each for $382.00
Only null left
Add to Cart
 
Description

Description

RCOR1/CoREST Polyclonal specifically detects RCOR1/CoREST in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

RCOR1/CoREST
Polyclonal
Rabbit
Transcription Factors and Regulators
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
KIAA0071CORESTRCORREST corepressor, Protein CoREST, REST corepressor 1
RCOR1
IgG
Affinity Purified
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human
Q9UKL0
23186
This antibody was developed against a recombinant protein corresponding to amino acids: SSDEEHGGGGMRVGPQYQAVVPDFDPAKLARRSQERDNLGMLV
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.