Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RCOR1/CoREST Antibody (SR2107), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321977100UL
Description
RCOR1/CoREST Monoclonal antibody specifically detects RCOR1/CoREST in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
RCOR1/CoREST | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence | |
SR2107 | |
Western Blot 1:1000-1:5000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
KIAA0071CORESTRCORREST corepressor, Protein CoREST, REST corepressor 1 | |
A synthesized peptide derived from human RCOR1/CoREST (Uniprot #: Q9UKL0) | |
100 μg | |
Transcription Factors and Regulators | |
23186 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction