Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RCOR1 Rabbit anti-Human, Polyclonal , Abnova™

Catalog No. 89934849 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-849 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-849 Supplier Abnova Corporation Supplier No. PAB29456
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant human RCOR1.

The RCOR gene encodes a functional corepressor required for regulation of neural-specific gene expression.[supplied by OMIM]

Sequence: SSDEEHGGGGMRVGPQYQAVVPDFDPAKLARRSQERDNLGMLV

Specifications

Antigen RCOR1
Applications Immunofluorescence, Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant human RCOR1.
Dilution Immunohistochemistry (1:20-1:50) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide)
Gene Alias COREST/KIAA0071/RCOR
Gene Symbols RCOR1
Host Species Rabbit
Immunogen Recombinant protein corresponding to human RCOR1.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23186
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.