Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RCOR3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5140042
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: RSRTSLMDRQ ARKLANRHNQ GDSDDDVEET HPMDGNDSDY DPKKEAKKEG NTEQPVQTSK IGLGRREYQS LQHRHHSQRS KCRPPKGMYL TQEDVVAVSC SPNAANT.
May act as a component of a corepressor complex that represses transcription. [UniProt].
Specifications
| RCOR3 | |
| Polyclonal | |
| Unconjugated | |
| Rcor3 | |
| 4921514E24Rik; C730034D20Rik; E130101E15Rik; FLJ10876; KIAA1343; Rcor3; REST corepressor 3; REST corepressor 3-like protein; si:ch211-284e20.6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55758 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9P2K3 | |
| Rcor3 | |
| Recombinant protein corresponding to Human RCOR3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction