Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RD Plastics Color Coded Ziplock Biohazard Specimen Bags
SDP

Catalog No. 23700202
Click to view available options
Color:
Green
Pink
Purple
Red

Color-coded 6 x 9 in. bags for easy identification. Available in green, red, pink or purple.

Color coded biohazard/specimen transport bag with tear top and paperwork pouch
  • 2 mil clear reclosable bag with tear-away top for easy opening
  • 3-wall construction for durable, reliable protection
  • Biohazard symbol printed in black ink on colored block

Specifications

Autoclavable Not Autoclavable
Capacity (English) 1.5 lb.
Closure Type Zip Seal
Color Green
For Use With (Application) Specimen transport
Includes Paperwork pouch
Material LDPE
No. per Case 1000
No. per Pack 100
Dimensions (L x W) 22.86 x 15.25cm
Length (English) 9 in.
Width (English) 6 in.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.