Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RD Plastics LABTITE™ Tranport Bag

Click to view available options
Dimensions (L x W):
10 x 6 in.
18 x 13 in.
Description
- Pull away release liner
- Press to close liner
- No glove-to adhesive contact
- Three wall construction
- Liquid-tight closure
- Designed polymer film
Specifications
Specifications
Color | Clear |
For Use With (Application) | Used for specimen transport |
Includes | Front pouch for requisition slip and separate back pouch, pull away release liner |
Thickness (Metric) | 0.279 mm |
Thickness (English) | 2 mil |
Dimensions (L x W) | 10 x 6 in. |
Length (English) | 10 in. |
Length (Metric) | 25.4 cm |
Product Type | Biohazard Specimen Transport Bag |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction