Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RD3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | RD3 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
RD3 Polyclonal specifically detects RD3 in Human samples. It is validated for Western Blot.Specifications
| RD3 | |
| Polyclonal | |
| Rabbit | |
| Q7Z3Z2 | |
| 343035 | |
| Synthetic peptides corresponding to RD3(retinal degeneration 3) The peptide sequence was selected from the N terminal of RD3. Peptide sequence GQMREAERQQRERSNAVRKVCTGVDYSWLASTPRSTYDLSPIERLQLEDV. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| LCA12C1orf36chromosome 1 open reading frame 36, protein RD3, retinal degeneration 3 | |
| RD3 | |
| IgG | |
| 23 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title