Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RD3 Polyclonal Antibody
RD3 Polyclonal Antibody
Supplier: Thermo Scientific PA583117
Description
RD3 Polyclonal Antibody for IHC (P)

Specifications
RD3 | |
Polyclonal | |
Unconjugated | |
RD3 | |
3322402L07Rik; C1orf36; LCA12; Protein RD3; RD3; rd-3; retinal degeneration 3; retinal degeneration 3, GUCY2D regulator; retinal degeneration protein 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
343035 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z3Z2 | |
RD3 | |
Recombinant protein corresponding to Human RD3. Recombinant protein control fragment (Product #RP-108328). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction