Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598582
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598582 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598582 Supplier Invitrogen™ Supplier No. PA598582
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RD3 Polyclonal Antibody for Western Blot, IHC (P), ELISA

RD3 is preferentially expressed in retina.Defects in RD3 are the cause of Leber congenital amaurosis type 12 (LCA12).
TRUSTED_SUSTAINABILITY

Specifications

Antigen RD3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene RD3
Gene Accession No. Q7Z3Z2, Q8BRE0
Gene Alias 3322402L07Rik; C1orf36; LCA12; Protein RD3; RD3; rd-3; retinal degeneration 3; retinal degeneration 3, GUCY2D regulator; retinal degeneration protein 3
Gene Symbols RD3
Host Species Rabbit
Immunogen Recombinant Human Protein RD3 protein (1-195AA).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 343035, 74023
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.