Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RDBP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565441
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565441 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565441 Supplier Invitrogen™ Supplier No. PA565441
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VYGEDMTPTLL RGAFSPFGNI IDLSMDPPRN CAFVTYEKME SADQAVAELN GTQVESVQLK VNIARKQPML DAATGKSVWG SLAVQNSPKG CHRDKRTQIV YSDDVYKE Highest antigen sequence identity to the following orthologs - mouse 96%, rat 96%.

RDBP is part of a complex termed negative elongation factor (NELF) which represses RNA polymerase II transcript elongation. This protein bears similarity to nuclear RNA-binding proteins; however, it has not been demonstrated that this protein binds RNA. The protein contains a tract of alternating basic and acidic residues, largely arginine (R) and aspartic acid (D). The protein encoded by this gene is part of a complex termed negative elongation factor (NELF) which represses RNA polymerase II transcript elongation. This protein bears similarity to nuclear RNA-binding proteins; however, it has not been demonstrated that this protein binds RNA. The protein contains a tract of alternating basic and acidic residues, largely arginine (R) and aspartic acid (D). The gene localizes to the major histocompatibility complex (MHC) class III region on chromosome 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RDBP
Applications ChIP Assay, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NELFE
Gene Accession No. P18615
Gene Alias D17H6S45; D6S45; major histocompatibility complex gene RD; negative elongation factor complex member E; negative elongation factor complex member E, Rdbp; negative elongation factor E; negative elongation factor polypeptide E; NELF E; Nelfe; NELF-E; nuclear protein; Rd; RD RNA binding protein; RD RNA-binding protein; RDBP; RDP; RNA-binding protein RD
Gene Symbols NELFE
Host Species Rabbit
Immunogen Recombinant Human RDBP. Recombinant protein control fragment (Product #RP-104821).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7936
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.