Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RDH10 Polyclonal Antibody
RDH10 Polyclonal Antibody
Supplier: Thermo Scientific PA565549
Description
RDH10 Polyclonal Antibody for ICC/IF

Specifications
RDH10 | |
Polyclonal | |
Unconjugated | |
RDH10 | |
3110069K09Rik; 4921506A21Rik; AI875664; AW549993; D1Ertd762e; m366Asp; RDH10; Retinol dehydrogenase 10; retinol dehydrogenase 10 (all-trans); SDR16C4; Short chain dehydrogenase/reductase family 16C member 4; short chain dehydrogenase/reductase family 16C, member 4; UNQ9375/PRO34191 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
157506 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IZV5 | |
RDH10 | |
Recombinant Human RDH10. Recombinant protein control fragment (Product #RP-106221). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction