Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RDH10 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5106301

Catalog No. PIPA5106301


Only null left
Add to Cart

Description

Description

Antibody detects endogenous levels of total RDH10.

RDH10 generates all-trans retinal from all-trans retinol and may plan an important role in the photic visual cycle. All-trans retinal is isomerized to 11-cis retinal by the retinal G protein-coupled receptor (RGR; MIM 600342) when the retinal pigment epithelium (RPE) is illuminated.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RDH10
Polyclonal
Unconjugated
RDH10
3110069K09Rik; 4921506A21Rik; AI875664; AW549993; D1Ertd762e; m366Asp; RDH10; Retinol dehydrogenase 10; retinol dehydrogenase 10 (all-trans); SDR16C4; Short chain dehydrogenase/reductase family 16C member 4; short chain dehydrogenase/reductase family 16C, member 4; UNQ9375/PRO34191
Rabbit
Affinity chromatography
RUO
157506, 353252, 98711
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q80ZF7, Q8IZV5, Q8VCH7
RDH10
A synthesized peptide derived from human RDH10(Accession Q8IZV5), corresponding to amino acid residues C242-L292.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.