Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RDH10 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981803
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1803 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1803 Supplier Bioss Supplier No. BS11941R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RDH10 generates all-trans retinal from all-trans retinol and may plan an important role in the photic visual cycle. All-trans retinal is isomerized to 11-cis retinal by the retinal G protein-coupled receptor (RGR; MIM 600342) when the retinal pigment epithelium (RPE) is illuminated.

Specifications

Antigen RDH10
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RDH10
Gene Accession No. Q8IZV5
Gene Alias 3110069K09Rik; 4921506A21Rik; AI875664; AW549993; BOS_13912; D1Ertd762e; m366Asp; RDH10; Retinol dehydrogenase 10; retinol dehydrogenase 10 (all-trans); SDR16C4; Short chain dehydrogenase/reductase family 16C member 4; short chain dehydrogenase/reductase family 16C, member 4; UNQ9375/PRO34191
Gene Symbols RDH10
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human RDH10.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 157506
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.