Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RDH11 Antibody (OTI1B4), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273843PE
Description
RDH11 Monoclonal specifically detects RDH11 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
RDH11 | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
RDH11 | |
Full length recombinant protein of human RDH11 (NP_057110) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG2a |
Flow Cytometry | |
OTI1B4 | |
Flow Cytometry | |
Androgen-regulated short-chain dehydrogenase/reductase 1, ARSDR1CGI82, EC 1.1.1.300, HCBP12, HCV core-binding protein HCBP12, MDT1, prostate short-chain dehydrogenase reductase 1, Prostate short-chain dehydrogenase/reductase 1, PSDR1FLJ32633, RALR1, Retinal reductase 1, retinol dehydrogenase 11, retinol dehydrogenase 11 (all-trans and 9-cis), retinol dehydrogenase 11 (all-trans/9-cis/11-cis), SCALD, SDR7C1, short chain dehydrogenase/reductase family 7C, member 1 | |
Mouse | |
Affinity Purified | |
Vision | |
51109.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction