Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RDH11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA573024
Description
RDH11 Polyclonal Antibody for Western Blot, IHC (F), IHC (P), IP
RHD11, a member of the short-chain dehydrogenase/reductase superfamily of oxidoreductases, is expressed at high levels in prostate epithelium, and its expression is regulated by androgens.
Specifications
RDH11 | |
Polyclonal | |
Unconjugated | |
Rdh11 | |
2610319N22Rik; AI428145; all-trans and 9-cis; androgen-regulated short-chain dehydrogenase/reductase 1; Arsdr1; AU045252; C85936; cell line MC/9.IL4 derived transcript 1; cell line MC/9.IL4-derived protein 1; CGI82; CGI-82; HCBP12; HCV core-binding protein HCBP12; M42C60; Mdt1; prostate short-chain dehydrogenase reductase 1; prostate short-chain dehydrogenase/reductase 1; Psdr1; ralR1; Rdh11; RDJCSS; Retinal reductase 1; retinol dehydrogenase 11; retinol dehydrogenase 11 (all-trans/9-cis/11-cis); SCALD; SDR7C1; Short chain dehydrogenase/reductase family 7C member 1; short chain dehydrogenase/reductase family 7C, member 1; short-chain aldehyde dehydrogenase; short-chain dehydrogenase/reductase 1; Ube-1c; UBE-1c1; ubiquitously expressing | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51109 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q8TC12 | |
Rdh11 | |
Synthetic peptides corresponding to the N terminal sequence MVELMFP and the C terminal sequence LLGLPID of the human RDH11/ PSDR1 protein. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction