Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RDH11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112685
Description
RDH11 Polyclonal Antibody for Western Blot, IHC (P), ELISA
RHD11, a member of the short-chain dehydrogenase/reductase superfamily of oxidoreductases, is expressed at high levels in prostate epithelium, and its expression is regulated by androgens.
Specifications
| RDH11 | |
| Polyclonal | |
| Unconjugated | |
| Rdh11 | |
| 2610319N22Rik; AI428145; all-trans and 9-cis; androgen-regulated short-chain dehydrogenase/reductase 1; Arsdr1; AU045252; C85936; cell line MC/9.IL4 derived transcript 1; cell line MC/9.IL4-derived protein 1; CGI82; CGI-82; HCBP12; HCV core-binding protein HCBP12; M42C60; Mdt1; prostate short-chain dehydrogenase reductase 1; prostate short-chain dehydrogenase/reductase 1; Psdr1; ralR1; Rdh11; RDJCSS; Retinal reductase 1; retinol dehydrogenase 11; retinol dehydrogenase 11 (all-trans/9-cis/11-cis); SCALD; SDR7C1; Short chain dehydrogenase/reductase family 7C member 1; short chain dehydrogenase/reductase family 7C, member 1; short-chain aldehyde dehydrogenase; short-chain dehydrogenase/reductase 1; Ube-1c; UBE-1c1; ubiquitously expressing | |
| Rabbit | |
| Protein G | |
| RUO | |
| 51109 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 4 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8TC12 | |
| Rdh11 | |
| Recombinant Human Retinol dehydrogenase 11 protein (22-318AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction