Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RDH11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501726855 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-6855 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-6855 Supplier Proteintech Group Inc Supplier No. 110771AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RHD11, a member of the short-chain dehydrogenase/reductase superfamily of oxidoreductases, is expressed at high levels in prostate epithelium, and its expression is regulated by androgens.

Specifications

Antigen RDH11
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene RDH11
Gene Accession No. Q8TC12, Q9QYF1
Gene Alias ARSDR1, CGI 82, HCBP12, MDT1, PSDR1, RALR1, RDH11, Retinal reductase 1, Retinol dehydrogenase 11, SCALD, SDR7C1
Gene Symbols Rdh11
Host Species Rabbit
Immunogen RDH11 Fusion Protein Ag1485
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17252, 362757, 51109
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.