Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RDH11 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987129
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7129 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7129 Supplier Bioss Supplier No. BS6214R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RHD11, a member of the short-chain dehydrogenase/reductase superfamily of oxidoreductases, is expressed at high levels in prostate epithelium, and its expression is regulated by androgens.

Specifications

Antigen RDH11
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Rdh11
Gene Accession No. Q8TC12
Gene Alias 2610319N22Rik; AI428145; all-trans and 9-cis; Androgen-regulated short-chain dehydrogenase/reductase 1; ARSDR1; AU045252; C85936; cell line MC/9.IL4 derived transcript 1; Cell line MC/9.IL4-derived protein 1; CGI82; CGI-82; HCBP12; HCV core-binding protein HCBP12; LOC100353716; M42C60; Mdt1; prostate short-chain dehydrogenase reductase 1; Prostate short-chain dehydrogenase/reductase 1; PSDR1; RalR1; Rdh11; RDJCSS; Retinal reductase 1; Retinol dehydrogenase 11; retinol dehydrogenase 11 (all-trans/9-cis/11-cis); SCALD; SDR7C1; Short chain dehydrogenase/reductase family 7C member 1; short chain dehydrogenase/reductase family 7C, member 1; Short-chain aldehyde dehydrogenase; short-chain dehydrogenase/reductase 1; Ube-1c; UBE-1c1; ubiquitously expressing
Gene Symbols Rdh11
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human RDH11.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51109
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.