Learn More
Invitrogen™ RDH12 Monoclonal Antibody (1E11)

Description
Peptide Sequence: RLQGTGVTTY AVHPGVVRSE LVRHSSLLCL LWRLFSPFVK TAREGAQTSL HCALAEGLEP LSGKYFSDCK RTWVSPRARN NKTAERLWNV SCELLGIRWE.
Specifications
Specifications
| Antigen | RDH12 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 1E11 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | RDH12 |
| Gene Accession No. | Q96NR8 |
| Gene Alias | A930033N07Rik; All-trans and 9-cis retinol dehydrogenase; double substrate specificity short-chain dehydrogenase/reductase 2; double substrate-specificity short chain dehydrogenase/reductase 2; DSSDR2; ds-SDR2; LCA13; LCA3; Rdh12; Retinol dehydrogenase 12; retinol dehydrogenase 12 (all-trans and 9-cis); retinol dehydrogenase 12 (all-trans/9-cis/11-cis); retinol dehydrogenase 12, all-trans and 9-cis; RP53; SDR7C2; short chain dehydrogenase/reductase family 7C member 2; short chain dehydrogenase/reductase family 7C, member 2; wu:fj43a10; zgc:92430 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.