Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RDH13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559818
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559818 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559818 Supplier Invitrogen™ Supplier No. PA559818
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GSGVTVNALH PGVARTELGR HTGIHGSTFS STTLGPIFWL LVKSPELAAQ PSTYLAVAEE LADVSGKYFD GL Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 83%.

Does not exhibit retinol dehydrogenase (RDH) activity in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RDH13
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rdh13
Gene Accession No. Q8NBN7
Gene Alias 8430425D21Rik; PSEC0082; Rdh13; Retinol dehydrogenase 13; retinol dehydrogenase 13 (all-trans and 9-cis); retinol dehydrogenase 13 (all-trans/9-cis); SDR7C3; Short chain dehydrogenase/reductase family 7C member 3; short chain dehydrogenase/reductase family 7C, member 3; UNQ736/PRO1430
Gene Symbols Rdh13
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RDH13. Recombinant protein control fragment (Product #RP-99103).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112724
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.