Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RDH14 Antibody (OTI1A7), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273847FR
Description
RDH14 Monoclonal specifically detects RDH14 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.Specifications
RDH14 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
RDH14 | |
Full length recombinant protein of human RDH14 (NP_065956) produced in HEK293T cells. | |
0.1 mL | |
57665.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry | |
OTI1A7 | |
Western Blot, Flow Cytometry | |
Alcohol dehydrogenase PAN2, EC 1.1.1.-, PAN2retinol dehydrogenase 14, pancreas protein 2, retinol dehydrogenase 14 (all-trans and 9-cis), retinol dehydrogenase 14 (all-trans/9-cis/11-cis), SDR7C4, short chain dehydrogenase/reductase family 7C, member 4 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction