Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RDH14 Monoclonal Antibody (OTI1A7)
GREENER_CHOICE

Catalog No. PIMA525430
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525430 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525430 Supplier Invitrogen™ Supplier No. MA525430
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RDH14 Monoclonal Antibody for Western Blot, Flow

Retinol dehydrogenase with a clear preference for NADP. Displays high activity towards 9-cis, 11-cis and all-trans-retinol. Shows a very weak activity towards 13-cis-retinol. Has no activity towards steroid. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RDH14
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI1A7
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Rdh14
Gene Accession No. Q9HBH5
Gene Alias 3110030G19Rik; Alcohol dehydrogenase PAN2; alcohol dehydrogenase PAN2; expressed sequence AW215439; AW215439; Pan2; pancreas protein 2; Rdh14; Retinol dehydrogenase 14; retinol dehydrogenase 14 (all-trans and 9-cis); retinol dehydrogenase 14 (all-trans and 9-cis)9; retinol dehydrogenase 14 (all-trans/9-cis/11-cis); RGD1565196; SDR7C4; short chain dehydrogenase/reductase family 7C member 4; short chain dehydrogenase/reductase family 7C, member 4; UNQ529/PRO1072
Gene Symbols Rdh14
Host Species Mouse
Immunogen Full length human recombinant protein of RDH14 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 500629, 57665
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.