Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RDH14 Monoclonal Antibody (OTI4G8), TrueMAB™, OriGene
SDP

Catalog No. 502541600
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1600 30 μL
50-167-3427 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1600 Supplier OriGene Supplier No. TA502365S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RDH14 Monoclonal antibody specifically detects RDH14 in Human samples. It is validated for Western Blot

Specifications

Antigen RDH14
Applications Western Blot
Classification Monoclonal
Clone OTI4G8
Concentration 0.83 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene OGFOD1
Gene Accession No. Q8N543
Gene Alias 3110030G19Rik; Alcohol dehydrogenase PAN2; alcohol dehydrogenase PAN2; expressed sequence AW215439; AW215439; Pan2; pancreas protein 2; Rdh14; Retinol dehydrogenase 14; retinol dehydrogenase 14 (all-trans and 9-cis); retinol dehydrogenase 14 (all-trans and 9-cis)9; retinol dehydrogenase 14 (all-trans/9-cis/11-cis); RGD1565196; SDR7C4; short chain dehydrogenase/reductase family 7C member 4; short chain dehydrogenase/reductase family 7C, member 4; UNQ529/PRO1072
Host Species Mouse
Immunogen Full length human recombinant protein of human OGFOD1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55239
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.