Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RDH14 Monoclonal Antibody (OTI6C3), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502315S


Description
RDH14 Monoclonal antibody specifically detects RDH14 in Canine, Human, Rat samples. It is validated for Western BlotSpecifications
RDH14 | |
Monoclonal | |
0.52 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q5VSL9 | |
Mouse | |
Affinity chromatography | |
RUO | |
362012, 490117, 85369 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Western Blot | |
OTI6C3 | |
Unconjugated | |
STRIP1 | |
3110030G19Rik; Alcohol dehydrogenase PAN2; alcohol dehydrogenase PAN2; expressed sequence AW215439; AW215439; Pan2; pancreas protein 2; Rdh14; Retinol dehydrogenase 14; retinol dehydrogenase 14 (all-trans and 9-cis); retinol dehydrogenase 14 (all-trans and 9-cis)9; retinol dehydrogenase 14 (all-trans/9-cis/11-cis); RGD1565196; SDR7C4; short chain dehydrogenase/reductase family 7C member 4; short chain dehydrogenase/reductase family 7C, member 4; UNQ529/PRO1072 | |
Full length human recombinant protein of human FAM40A produced in HEK293T cell. | |
30 μL | |
Primary | |
Canine, Human, Rat | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction