Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RDH16 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558423
Description
Immunogen sequence: GVKVAMIEPG YFKTAVTSKE RFLKSFLEIW DRSSPEVKEA YGEKFVADYK KSAEQMEQKC TQDLSLVT Highest antigen sequence identity to the following orthologs: Mouse - 54%, Rat - 54%.
Retinol dehydrogenase 16 (RDH16) is an oxidoreductase with a preference for NAD. It oxidizes all-trans-retinol and 13-cis-retinol to the corresponding aldehydes. RDH16 has higher activity towards CRBP-bound retinol than with free retinol. It oxidizes 3-alpha-hydroxysteroids and also oxidizes androstanediol and androsterone to dihydrotestosterone and androstanedione. RDH16 can also catalyze the reverse reaction.
Specifications
RDH16 | |
Polyclonal | |
Unconjugated | |
RDH16 | |
29 kDa protein; Cis-retinol androgen dehydrogenase 1; cis-retinol/androgen dehydrogenase 1; Crad; CRAD1; hRDH-E; Human epidermal retinol dehydrogenase; Microsomal NAD(+)-dependent retinol dehydrogenase 4; microsomal NAD+-dependent retinol dehydrogenase 4; Rdh1; Rdh16; Rdh2; Rdh6; Retinol dehydrogenase 16; retinol dehydrogenase 16 (all-trans and 13-cis); retinol dehydrogenase 16 (all-trans); retinol dehydrogenase 2; retinol dehydrogenase 6; Retinol dehydrogenase type 1; retinol dehydrogenase type 2; retinol dehydrogenase type 6; Retinol dehydrogenase type II; retinol dehydrogenase type II (RODH II); RODH II; Rodh1; RODH4; RoDH-4; Rodhii; SDR9C8; Short chain dehydrogenase/reductase family 9C member 8; short chain dehydrogenase/reductase family 9C, member 8; sterol/retinol dehydrogenase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8608 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.08 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75452 | |
RDH16 | |
Recombinant protein corresponding to Human RDH16. Recombinant protein control fragment (Product #RP-97767). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction