Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RDH5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530150
Description
Recommended positive controls: A549, HeLa, Jurkat, NT2D1. Predicted reactivity: Mouse (89%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes an enzyme belonging to the short-chain dehydrogenases/reductases family. This retinol dehydrogenase functions to catalyze the final step in the biosynthesis of 11-cis retinaldehyde, which is the universal chromophore of visual pigments. Mutations in this gene cause autosomal recessive fundus albipunctatus, a rare form of night blindness that is characterized by a delay in the regeneration of cone and rod photopigments. Alternative splicing results in multiple transcript variants. Read-through transcription also exists between this gene and the neighboring upstream BLOC1S1 gene.
Specifications
RDH5 | |
Polyclonal | |
Unconjugated | |
RDH5 | |
11-cis RDH; 11-cis retinol dehydrogenase; 11-cis RoDH; 9-cis; 9-cis retinol dehydrogenase; 9-cis-retinol dehydrogenase; 9-cis-retinol specific dehydrogenase; 9cRDH; AI987873; Cis-retinol dehydrogenase; cRDH; FLJ39337; FLJ97089; HSD17B9; rdh; RDH1; Rdh4; rdh5; retinol dehydrogenase 1; retinol dehydrogenase 5; retinol dehydrogenase 5 (11-cis and 9-cis); retinol dehydrogenase 5 (11-cis/9-cis); retinol dehydrogenase type 5; SDR9C5; Short chain dehydrogenase/reductase family 9C member 5; short chain dehydrogenase/reductase family 9C, member 5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5959 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q92781 | |
RDH5 | |
Recombinant fragment corresponding to a region within amino acids 60 and 276 of Human RDH5. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction