Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RDH5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566575
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566575 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566575 Supplier Invitrogen™ Supplier No. PA566575
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NNAGVAGIIGP TPWLTRDDFQ RVLNVNTMGP IGVTLALL Highest antigen sequence identity to the following orthologs - mouse 92%, rat 95%.

This gene encodes an enzyme belonging to the short-chain dehydrogenases/reductases family. This retinol dehydrogenase functions to catalyze the final step in the biosynthesis of 11-cis retinaldehyde, which is the universal chromophore of visual pigments. Mutations in this gene cause autosomal recessive fundus albipunctatus, a rare form of night blindness that is characterized by a delay in the regeneration of cone and rod photopigments. Alternative splicing results in multiple transcript variants. Read-through transcription also exists between this gene and the neighboring upstream BLOC1S1 gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RDH5
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RDH5
Gene Accession No. Q92781
Gene Alias 11-cis RDH; 11-cis retinol dehydrogenase; 11-cis RoDH; 9-cis; 9-cis retinol dehydrogenase; 9-cis-retinol dehydrogenase; 9-cis-retinol specific dehydrogenase; 9cRDH; AI987873; Cis-retinol dehydrogenase; cRDH; FLJ39337; FLJ97089; HSD17B9; rdh; RDH1; Rdh4; rdh5; retinol dehydrogenase 1; retinol dehydrogenase 5; retinol dehydrogenase 5 (11-cis and 9-cis); retinol dehydrogenase 5 (11-cis/9-cis); retinol dehydrogenase type 5; SDR9C5; Short chain dehydrogenase/reductase family 9C member 5; short chain dehydrogenase/reductase family 9C, member 5
Gene Symbols RDH5
Host Species Rabbit
Immunogen Recombinant Human RDH5. Recombinant protein control fragment (Product #RP-107230).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5959
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.