Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RDH5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519319
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519319 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519319 Supplier Invitrogen™ Supplier No. PA519319
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.

This gene encodes an enzyme belonging to the short-chain dehydrogenases/reductases family. This retinol dehydrogenase functions to catalyze the final step in the biosynthesis of 11-cis retinaldehyde, which is the universal chromophore of visual pigments. Mutations in this gene cause autosomal recessive fundus albipunctatus, a rare form of night blindness that is characterized by a delay in the regeneration of cone and rod photopigments. Alternative splicing results in multiple transcript variants. Read-through transcription also exists between this gene and the neighboring upstream BLOC1S1 gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RDH5
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene RDH5
Gene Accession No. Q92781
Gene Alias 11-cis RDH; 11-cis retinol dehydrogenase; 11-cis RoDH; 9-cis; 9-cis retinol dehydrogenase; 9-cis-retinol dehydrogenase; 9-cis-retinol specific dehydrogenase; 9cRDH; AI987873; Cis-retinol dehydrogenase; cRDH; FLJ39337; FLJ97089; HSD17B9; rdh; RDH1; Rdh4; rdh5; retinol dehydrogenase 1; retinol dehydrogenase 5; retinol dehydrogenase 5 (11-cis and 9-cis); retinol dehydrogenase 5 (11-cis/9-cis); retinol dehydrogenase type 5; SDR9C5; Short chain dehydrogenase/reductase family 9C member 5; short chain dehydrogenase/reductase family 9C, member 5
Gene Symbols RDH5
Host Species Goat
Immunogen Synthetic peptide sequence (RDRQSLPASNAFV) corresponding to the internal amino acids of RDH5 (aa 19-31).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5959
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.