Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RDH8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570685
Description
This target displays homology in the following species: Cow: 93%; Dog: 92%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.
Retinol dehydrogenase with a clear preference for NADP. Converts all-trans-retinal to all-trans-retinol. May play a role in the regeneration of visual pigment at high light intensity.
Specifications
RDH8 | |
Polyclonal | |
Unconjugated | |
RDH8 | |
Gm182; LOW QUALITY PROTEIN: retinol dehydrogenase 8; photoreceptor outer segment all-trans retinol dehydrogenase; PRRDH; Rdh8; Retinol dehydrogenase 8; retinol dehydrogenase 8 (all-trans); SDR28C2; Short chain dehydrogenase/reductase family 28C member 2; short chain dehydrogenase/reductase family 28C, member 2; visual cycle enzyme that reduces all-trans retinal to all-trans retinol | |
Rabbit | |
Affinity chromatography | |
RUO | |
50700 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NYR8 | |
RDH8 | |
synthetic peptide directed towards the middle region of human RDH8. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction