Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RDH8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5139867
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: VQAIVNVISS TRPPLRRQTN IRYSPLTTLK TVDSSGSLYV RTTHRLLFRC PRLLNLGLQC L.
Retinol dehydrogenase with a clear preference for NADP. Converts all-trans-retinal to all-trans-retinol. May play a role in the regeneration of visual pigment at high light intensity.
Specifications
RDH8 | |
Polyclonal | |
Unconjugated | |
RDH8 | |
Gm182; LOW QUALITY PROTEIN: retinol dehydrogenase 8; photoreceptor outer segment all-trans retinol dehydrogenase; PRRDH; Rdh8; Retinol dehydrogenase 8; retinol dehydrogenase 8 (all-trans); SDR28C2; Short chain dehydrogenase/reductase family 28C member 2; short chain dehydrogenase/reductase family 28C, member 2; visual cycle enzyme that reduces all-trans retinal to all-trans retinol | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
50700 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NYR8 | |
RDH8 | |
Recombinant protein corresponding to Human RDH8. Recombinant protein control fragment (Product #RP-109062). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction