Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RDH8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571449
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571449 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571449 Supplier Invitrogen™ Supplier No. PA571449
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RDH8 Polyclonal Antibody for Western Blot

Retinol dehydrogenase with a clear preference for NADP. Converts all-trans-retinal to all-trans-retinol. May play a role in the regeneration of visual pigment at high light intensity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RDH8
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RDH8
Gene Accession No. Q9NYR8
Gene Alias Gm182; LOW QUALITY PROTEIN: retinol dehydrogenase 8; photoreceptor outer segment all-trans retinol dehydrogenase; PRRDH; Rdh8; Retinol dehydrogenase 8; retinol dehydrogenase 8 (all-trans); SDR28C2; Short chain dehydrogenase/reductase family 28C member 2; short chain dehydrogenase/reductase family 28C, member 2; visual cycle enzyme that reduces all-trans retinal to all-trans retinol
Gene Symbols RDH8
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human RDH8.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50700
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.