Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RDM1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen RDM1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP153147
SDP
View Documents
Novus Biologicals
NBP153147
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

RDM1 Polyclonal specifically detects RDM1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

RDM1
Polyclonal
Rabbit
A8MY68
201299
Synthetic peptides corresponding to RDM1(RAD52 motif 1) The peptide sequence was selected from the middle region of RDM1. Peptide sequence NSSKCQELANYYFGFNGCSKRIIKLQELSDLEERENEDSMVPLPKQSLKF.
Primary
Western Blot
Unconjugated
RUO
MGC33977, RAD52 homolog B, RAD52 homolog B (S. cerevisiae), RAD52 motif 1, RAD52 motif-containing protein 1, RAD52B
RDM1
IgG
26 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.