Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RDM1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110906
Description
Immunogen sequence: SPVKVRLGTR HKAVQHQALA LNSSKCQELA NYYFGFNGCS KRIIKLQELS DLEERENEDS MVPLPKQSLK FFCALEVVLP SCDCRSPGIG LVE.
This gene encodes a protein involved in the cellular response to cisplatin, a drug commonly used in chemotherapy. The protein encoded by this gene contains two motifs: a motif found in RAD52, a protein that functions in DNA double-strand breaks and homologous recombination, and an RNA recognition motif (RRM) that is not found in RAD52. The RAD52 motif region in RAD52 is important for protein function and may be involved in DNA binding or oligomerization. Alternatively spliced transcript variants encoding different isoforms have been reported.
Specifications
RDM1 | |
Polyclonal | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NG50 | |
RDM1 | |
Recombinant Protein corresonding to Human RDM1. Recombinant protein control fragment (Product #RP-93528). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot, Immunocytochemistry | |
Unconjugated | |
RDM1 | |
2410008M22Rik; AW212028; DeltaN-RDM1zeta; RAD52 homolog B; RAD52 motif 1; RAD52 motif containing 1; RAD52 motif-containing protein 1; RAD52B; RAD52B homolog; Rdm1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
201299 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction