Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RDX Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501735212 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5212 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5212 Supplier Proteintech Group Inc Supplier No. 261051AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ezrin, radixin and moesin (ERM) proteins are widely distributed proteins located in the cellular cortex, in microvilli and adherens junctions. They are mediate linkage of actin cytoskeleton to plasma membrane in many cells. ERM proteins contain an N-terminal FERM (4.1/ezrin/radixin/moesin) domain that binds to phosphatidylinositol-(4, 5)phosphate and cellular membrane proteins. The ERM, particularly ezrin, is important for reconstructing cell-surface architecture during T cell activation. Despite the high degree of homology, the three proteins exhibit a distinct receptor-specific pattern of phosphorylation. ERM activity is regulated in part by phosphorylation at a C-terminal threonine (ezrin-Thr567, radixin-Thr564, moesin-Thr558).

Specifications

Antigen RDX
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.24 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene RDX
Gene Accession No. P26043, P35241
Gene Alias DFNB24, radixin, RDX
Gene Symbols RDX
Host Species Rabbit
Immunogen RDX Fusion Protein Ag23532
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19684, 5962
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.